Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008326.1 | NP_001008327.1 | 7 | 955 | ||
GenBank mRNA | BC086420.1 | - | 7 | 955 | 7300740 |