Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008343.1 | NP_001008344.1 | 6 | 1241 | ||
GenBank mRNA | BC085823.1 | - | 6 | 1241 | 7126835 |