Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008346.1 | NP_001008347.1 | 2 | 350 | ||
GenBank mRNA | BC085119.1 | - | 2 | 350 | 7108537 |