Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008350.1 | NP_001008351.1 | 2 | 384 | ||
XM_006256191.2 | XP_006256253.1 | 3 | 1214 | |||
XM_006256192.2 | XP_006256254.1 | 3 | 1015 | |||
XM_008772812.1 | XP_008771034.1 | 3 | 998 | |||
GenBank mRNA | BC086421.1 | - | 2 | 384 | 7300732 | |
FQ231020.1 | - | 1 | 79 |