Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008354.1 | NP_001008355.1 | 3 | 264 | ||
XM_006234068.1 | XP_006234130.1 | 3 | 298 | |||
GenBank mRNA | BC085779.1 | - | 3 | 264 | 7106905 |