Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata31a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008359.2 | NP_001008360.2 | 1, 2 | 221 | ||
GenBank mRNA | AF511042.2 | - | 1, 2 | 135 | ||
BC085936.1 | - | 1, 2 | 204 | 7115545 | ||
CK653072.1 | - | 1, 2 | 221 |