Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008361.1 | NP_001008362.1 | 4 | 959 | ||
XM_006246750.2 | XP_006246812.1 | 4 | 373 | |||
GenBank mRNA | BC083670.1 | - | 4 | 959 | 7135651 | |
FQ216790.1 | - | 4 | 293 |