Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008362.1 | NP_001008363.1 | 4 | 602 | ||
XM_006248895.2 | XP_006248957.1 | 4 | 696 | |||
XM_008768977.1 | XP_008767199.1 | 4 | 968 | |||
XM_008768978.1 | XP_008767200.1 | 4 | 1057 | |||
GenBank mRNA | BC085937.1 | - | 4 | 602 | 7121505 |
Gene Symbol: LOC103690102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769057.1 | XP_008767279.1 | 3 | 511 |