Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008373.1 | NP_001008374.1 | 1977 | |||
GenBank mRNA | AY236996.1 | - | 2018 | |||
BC086553.1 | - | 1977 | 7303262 |
Gene Symbol: LOC100362980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242128.2 | XP_006242190.1 | 2 | 2330 |