Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008378.1 | NP_001008379.1 | 6 | 1263 | ||
GenBank mRNA | BC086452.1 | - | 6 | 1263 | 7307024 | |
FQ214182.1 | - | 6 | 1234 | |||
FQ218380.1 | - | 6 | 1234 | |||
FQ219469.1 | - | 6 | 1258 | |||
FQ219661.1 | - | 6 | 1262 |