Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008394.2 | NP_001008395.1 | 1 | 1186 | ||
GenBank mRNA | AK098698.1 | - | 1 | 1179 | ||
BC027612.2 | - | 1 | 1147 | 4831023 |