Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008397.3 | NP_001008398.2 | 3 | 950 | ||
NM_001306197.1 | NP_001293126.1 | 3 | 797 | |||
NM_001306198.1 | NP_001293127.1 | 2 | 688 | |||
NM_001306201.1 | NP_001293130.1 | 3 | 671 | |||
NR_131336.1 | 4 | 864 | ||||
NR_131337.1 | 4 | 806 | ||||
NR_131338.1 | 3 | 602 | ||||
NR_131340.1 | 3 | 940 | ||||
XM_006714631.2 | XP_006714694.1 | 3 | 916 | |||
GenBank mRNA | AK074216.1 | - | 3 | 953 | ||
AK298545.1 | - | 3 | 745 | |||
AK307365.1 | - | 2 | 626 | |||
AY358715.1 | - | 3 | 876 | |||
BC029424.1 | - | 3 | 909 | 4285958 | ||
BX537894.1 | - | 4 | 814 | |||
BX647770.1 | - | 2 | 638 |