Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008419.2 | NP_001008419.1 | 24, 25 | 2771 | ||
GenBank mRNA | AY187018.2 | - | 24, 25 | 2771 | ||
AY665589.1 | - | 24, 25 | 2678 | |||
BC150826.1 | - | 24, 25 | 2698 | 9087737 |