Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014698.2 | NP_055513.2 | 7 | 1061 | ||
XM_006711841.3 | XP_006711904.1 | 7 | 715 | |||
XM_011544328.2 | XP_011542630.1 | 7 | 913 | |||
XM_011544329.2 | XP_011542631.1 | 7 | 2180 | |||
XM_011544330.2 | XP_011542632.1 | 6 | 870 | |||
XM_011544331.2 | XP_011542633.1 | 7 | 744 | |||
XM_011544332.2 | XP_011542634.1 | 7 | 744 | |||
XM_017002916.1 | XP_016858405.1 | 7 | 715 | |||
GenBank mRNA | AB018335.1 | - | 7 | 689 | ||
AK022502.1 | - | 7 | 710 | |||
AK222934.1 | - | 7 | 691 | |||
AK222944.1 | - | 7 | 691 | |||
AK309210.1 | - | 5 | 638 | |||
BC030245.1 | - | 7 | 694 | 5216414 |