Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008537.2 | NP_001008537.1 | 4 | 6992 | ||
GenBank mRNA | AB095942.1 | - | 5 | 6620 | ||
AY563507.1 | - | 4 | 6689 | |||
BC152557.1 | - | 5 | 6620 | 8860127 |