Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014704.3 | NP_055519.1 | 6, 7 | 920 | ||
XM_005244815.4 | XP_005244872.1 | 7, 8 | 1020 | |||
XM_011542473.2 | XP_011540775.1 | 7, 8 | 1220 | |||
XM_011542474.2 | XP_011540776.1 | 6, 7 | 938 | |||
XM_011542475.2 | XP_011540777.1 | 7, 8 | 1220 | |||
XM_011542476.2 | XP_011540778.1 | 7, 8 | 1220 | |||
XM_011542477.2 | XP_011540779.1 | 7, 8 | 1220 | |||
XM_011542478.1 | XP_011540780.1 | 7, 8 | 1220 | |||
XM_017002917.1 | XP_016858406.1 | 7, 8 | 1202 | |||
XM_017002918.1 | XP_016858407.1 | 6, 7 | 920 | |||
XM_017002919.1 | XP_016858408.1 | 6, 7 | 898 | |||
GenBank mRNA | AB011134.1 | - | 6, 7 | 874 | ||
BC001640.2 | - | 6, 7 | 885 | |||
BC047450.1 | - | 6, 7 | 885 | 5297035 | ||
DA081362.1 | - | 4, 5 | 362 |