Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psenen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008764.2 | NP_001008764.1 | 4 | 332 | ||
XM_006228770.2 | XP_006228832.1 | 4 | 384 | |||
GenBank mRNA | AY539891.1 | - | 3 | 223 | ||
BC100104.1 | - | 4 | 332 | 7377803 |