Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008770.3 | NP_001008770.1 | 4 | 474 | ||
XM_006232097.2 | XP_006232159.1 | 4 | 2585 | |||
GenBank mRNA | AY325197.1 | - | 4 | 476 | ||
BC088459.1 | - | 3 | 458 | 7127481 | ||
BF556107.1 | - | 4 | 481 | |||
FQ209709.1 | - | 4 | 632 | |||
FQ213936.1 | - | 4 | 484 | |||
FQ215832.1 | - | 3 | 476 | |||
FQ219291.1 | - | 4 | 563 | |||
FQ219760.1 | - | 4 | 487 |