Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008773.2 | NP_001008773.2 | 3 | 777 | ||
XM_006254688.2 | XP_006254750.1 | 3 | 894 | |||
XM_006254689.1 | XP_006254751.1 | 2 | 632 | |||
GenBank mRNA | AY325156.1 | - | 2 | 401 | ||
BC093607.1 | - | 3 | 777 | 7318425 |