Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206938.1 | NP_001193867.1 | 2 | 520 | ||
NM_014719.2 | NP_055534.1 | 2 | 480 | |||
XM_005250074.3 | XP_005250131.1 | 2 | 520 | |||
XM_005250075.2 | XP_005250132.1 | 2 | 522 | |||
XM_005250076.3 | XP_005250133.1 | 2 | 557 | |||
XM_005250077.2 | XP_005250134.1 | 3 | 686 | |||
XM_006716191.2 | XP_006716254.1 | 2 | 486 | |||
XM_006716192.3 | XP_006716255.1 | 2 | 629 | |||
XM_017012833.1 | XP_016868322.1 | 2 | 552 | |||
XM_017012834.1 | XP_016868323.1 | 2 | 522 | |||
XM_017012835.1 | XP_016868324.1 | 3 | 686 | |||
XM_017012836.1 | XP_016868325.1 | 2 | 486 | |||
GenBank mRNA | AB018281.1 | - | 2 | 480 | ||
AK001640.1 | - | 2 | 556 | |||
AK291605.1 | - | 2 | 554 | |||
BC000609.2 | - | 2 | 461 | 3346754 | ||
DA196926.1 | - | 2 | 525 | |||
DA317873.1 | - | 2 | 520 |