Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008812.1 | NP_001008812.1 | 5 | 1031 | ||
XM_006242346.2 | XP_006242408.1 | 5 | 1100 |