Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097579.1 | NP_001091048.1 | 3 | 1044 | ||
NM_005300.3 | NP_005291.1 | 3 | 968 | |||
XM_005272597.3 | XP_005272654.1 | 3 | 3471 | |||
GenBank mRNA | AF039686.1 | - | 1 | 838 | ||
AK027780.1 | - | 3 | 1044 | |||
AK074627.1 | - | 3 | 1190 | |||
AK075217.1 | - | 3 | 959 | |||
AK122945.1 | - | 1 | 2399 | |||
BC020678.1 | - | 3 | 935 | 4770479 | ||
DQ106398.1 | - | 3 | 1035 | |||
DQ106399.1 | - | 5 | 1006 | |||
DQ106400.1 | - | 5 | 1052 | |||
DQ106401.1 | - | 3 | 894 | |||
DQ106402.1 | - | 4 | 896 |
Set Membership: |
G-protein coupled receptor |