Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014742.3 | NP_055557.2 | 4 | 516 | ||
XM_005260622.4 | XP_005260679.1 | 4 | 568 | |||
XM_017028154.1 | XP_016883643.1 | 4 | 297 | |||
XM_017028155.1 | XP_016883644.1 | 4 | 568 | |||
XM_017028159.1 | XP_016883648.1 | 4 | 349 | |||
XM_017028161.1 | XP_016883650.1 | 4 | 297 | |||
GenBank mRNA | AK296500.1 | - | 4 | 296 | ||
AK302439.1 | - | 4, 5 | 371 | |||
AK316293.1 | - | 3 | 220 | |||
BC021107.1 | - | 4 | 292 | 4896167 | ||
BC022850.2 | - | 3 | 274 | |||
D87444.1 | - | 4 | 570 | |||
DC381660.1 | - | 4 | 516 |