Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009268.1 | NP_001009268.1 | 4 | 496 | ||
GenBank mRNA | BC088442.1 | - | 4 | 496 | 7126325 | |
FQ212547.1 | - | 4 | 495 | |||
FQ212814.1 | - | 5 | 460 | |||
FQ219071.1 | - | 4 | 445 | |||
FQ219838.1 | - | 4 | 462 | |||
FQ225424.1 | - | 4 | 467 | |||
FQ226083.1 | - | 4 | 442 | |||
FQ226422.1 | - | 4 | 445 | |||
FQ227142.1 | - | 4 | 445 | |||
FQ227488.1 | - | 4 | 494 | |||
FQ227830.1 | - | 4 | 445 | |||
FQ230874.1 | - | 4 | 480 | |||
FQ231828.1 | - | 4 | 447 | |||
FQ231874.1 | - | 4 | 480 | |||
FQ234230.1 | - | 4 | 447 | |||
FQ235071.1 | - | 4 | 467 | |||
FQ235201.1 | - | 4 | 462 |