Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009283.1 | NP_001009283.1 | 2 | 246 | ||
XM_006228866.2 | XP_006228928.1 | 2 | 550 | |||
XM_006228867.1 | XP_006228929.1 | 2 | 337 | |||
XM_006228868.2 | XP_006228930.1 | 2 | 550 | |||
XM_006228869.2 | XP_006228931.1 | 2 | 557 | |||
XM_008759132.1 | XP_008757354.1 | 2 | 337 | |||
GenBank mRNA | BC088237.1 | - | 2 | 246 | 7325652 |
Gene Symbol: LOC100911963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748821.2 | XP_003748869.1 | 2 | 337 | ||
XM_003748822.2 | XP_003748870.1 | 2 | 337 | |||
XM_006228801.2 | XP_006228863.1 | 2 | 551 | |||
XM_006228802.2 | XP_006228864.1 | 2 | 551 | |||
XM_006228803.2 | XP_006228865.1 | 2 | 555 | |||
XM_008759243.1 | XP_008757465.1 | 2 | 337 |