Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014753.3 | NP_055568.3 | 5 | 980 | ||
XM_005271846.3 | XP_005271903.1 | 5 | 855 | |||
XM_005271848.3 | XP_005271905.1 | 5 | 711 | |||
XM_005271849.3 | XP_005271906.1 | 5 | 711 | |||
XM_011540402.2 | XP_011538704.1 | 5 | 711 | |||
GenBank mRNA | AK308420.1 | - | 5 | 687 | ||
BC043345.1 | - | 5 | 675 | 6062924 | ||
BC150252.1 | - | 4 | 649 | 9007274 | ||
D80009.1 | - | 4 | 649 |