Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009369.2 | NP_001009369.2 | 7 | 844 | ||
XM_008768372.1 | XP_008766594.1 | 7 | 1632 | |||
GenBank mRNA | BC087096.1 | - | 7 | 841 | 7191805 |