Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241250.2 | XP_006241312.1 | 3 | 866 | ||
GenBank mRNA | BC088210.1 | - | 3 | 770 | 7382451 |