Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009466.2 | NP_001009466.1 | 3 | 562 | ||
XM_006239647.2 | XP_006239709.1 | 3 | 664 | |||
GenBank mRNA | AY053516.1 | - | 3 | 563 | ||
BC158565.1 | - | 4 | 768 | |||
FQ209637.1 | - | 1 | 273 |