Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009466.2 | NP_001009466.1 | 3 | 587 | ||
GenBank mRNA | AY053516.1 | - | 3 | 588 |