Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009474.1 | NP_001009474.1 | 7 | 723 | ||
GenBank mRNA | BC088290.1 | - | 7 | 723 | 7322321 |