Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270975.1 | NP_001257904.1 | 3 | 234 | ||
NM_001270976.1 | NP_001257905.1 | 4 | 349 | |||
NM_001270977.1 | NP_001257906.1 | 3 | 234 | |||
NM_014761.3 | NP_055576.2 | 3 | 234 | |||
GenBank mRNA | AB097052.1 | - | 3 | 185 | ||
AK057902.1 | - | 3 | 197 | |||
AK291388.1 | - | 3 | 219 | |||
AK293022.1 | - | 3 | 195 | |||
AK293040.1 | - | 4 | 337 | |||
AK295066.1 | - | 3 | 196 | |||
AK297088.1 | - | 1 | 197 | |||
AK297336.1 | - | 3 | 217 | |||
BC000116.1 | - | 3 | 174 | 3350789 | ||
BC000430.2 | - | 3 | 184 | 2819828 | ||
BC004359.1 | - | 3 | 184 | 2819828 | ||
BC103745.1 | - | 3 | 212 | 6009274 | ||
D79996.1 | - | 3 | 185 | |||
DA698420.1 | - | 3 | 234 |