Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009504.1 | NP_001009504.1 | 5 | 1397 | ||
XM_006246020.2 | XP_006246082.1 | 6 | 1715 | |||
GenBank mRNA | AY823669.1 | - | 5 | 1397 |