Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009533.2 | NP_001009533.2 | 1 | 2081 | ||
GenBank mRNA | BC060520.1 | - | 2 | 2057 | 5597839 |