Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 17, 18 | 2668 | ||
NM_003703.2 | NP_003694.1 | 17, 18 | 2668 | |||
GenBank mRNA | AB000467.1 | - | 15, 16 | 2064 | ||
AF040965.1 | - | 17, 18 | 2530 | |||
AK024755.1 | - | 7, 8 | 977 | |||
AK025692.1 | - | 11, 12 | 1533 | |||
AK292939.1 | - | 17, 18 | 2668 | |||
BC053635.1 | - | 17, 18 | 2547 | 5114280 |