Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 7 | 1120 | ||
NM_001291979.1 | NP_001278908.1 | 7 | 1120 | |||
NM_003703.2 | NP_003694.1 | 7 | 1120 | |||
GenBank mRNA | AB000467.1 | - | 5 | 516 | ||
AF040965.1 | - | 7 | 982 | |||
AK292939.1 | - | 7 | 1120 | |||
BC009760.2 | - | 3 | 314 | |||
BC026035.1 | - | 7 | 947 | 4878317 | ||
BC053635.1 | - | 7 | 999 | 5114280 |