Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008255.2 | NP_032281.2 | 11 | 1481 | ||
GenBank mRNA | AK159899.1 | - | 10 | 1369 | ||
BC019782.1 | - | 10 | 1357 | |||
BC059873.1 | - | 2 | 162 | |||
BC085083.1 | - | 11 | 1393 | 5320486 |