Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304447.1 | NP_001291376.1 | 10 | 3695 | ||
NM_014653.3 | NP_055468.2 | 9 | 3864 | |||
XM_017020243.1 | XP_016875732.1 | 10 | 4149 | |||
XM_017020244.1 | XP_016875733.1 | 11 | 4310 | |||
XM_017020245.1 | XP_016875734.1 | 11 | 3807 | |||
XM_017020246.1 | XP_016875735.1 | 9 | 3538 | |||
XM_017020247.1 | XP_016875736.1 | 10 | 3681 | |||
XM_017020248.1 | XP_016875737.1 | 10 | 4153 | |||
XM_017020249.1 | XP_016875738.1 | 9 | 4010 | |||
XM_017020250.1 | XP_016875739.1 | 7 | 3564 | |||
XM_017020251.1 | XP_016875740.1 | 8 | 3375 | |||
XM_017020252.1 | XP_016875741.1 | 8 | 3853 | |||
GenBank mRNA | AB018332.2 | - | 9 | 3601 | ||
AK098069.1 | - | 1 | 1396 | |||
BC037969.1 | - | 5 | 2150 | |||
BC110330.1 | - | 9 | 3647 | 5735001 | ||
BC136687.1 | - | 8 | 3393 | 9020677 | ||
BC136688.1 | - | 8 | 3393 | 9020678 | ||
BX647283.1 | - | 10 | 3646 |