Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014784.3 | NP_055599.1 | 7 | 1596 | ||
NM_198236.2 | NP_937879.1 | 7 | 1596 | |||
XM_005245629.4 | XP_005245686.1 | 7 | 1997 | |||
XM_006711659.2 | XP_006711722.1 | 9 | 2045 | |||
XM_006711660.3 | XP_006711723.1 | 9 | 2045 | |||
XM_006711661.2 | XP_006711724.1 | 9 | 2045 | |||
XM_006711662.2 | XP_006711725.1 | 9 | 2045 | |||
XM_006711663.3 | XP_006711726.1 | 7 | 1032 | |||
XM_006711665.3 | XP_006711728.1 | 9 | 2045 | |||
XM_011510185.2 | XP_011508487.1 | 8 | 2030 | |||
XM_011510186.2 | XP_011508488.1 | 9 | 2045 | |||
XM_011510187.2 | XP_011508489.1 | 8 | 756 | |||
XM_017002920.1 | XP_016858409.1 | 8 | 2012 | |||
XM_017002921.1 | XP_016858410.1 | 7 | 1997 | |||
XM_017002922.1 | XP_016858411.1 | 9 | 2045 | |||
XM_017002923.1 | XP_016858412.1 | 7 | 1997 | |||
XM_017002924.1 | XP_016858413.1 | 8 | 2030 | |||
GenBank mRNA | AB002378.1 | - | 7 | 1301 | ||
BC057394.1 | - | 7 | 1620 | 6148684 |