Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LIMCH1 SIRNA Assays ›
Gene Symbol:
LIMCH1
Gene Name:
LIM and calponin homology domains 1
Gene Aliases:
LIMCH1A, LMO7B
Chromosome Location:
Chr.4: 41359607 - 41700044 on Build GRCh38
UniGene ID:
Hs.335163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
219453
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LIMCH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001112717.2
NP_001106188.1
20, 21
2615
NM_001112718.2
NP_001106189.1
20, 21
2612
NM_001112719.2
NP_001106190.1
15, 16
2397
NM_001112720.2
NP_001106191.1
14, 15
2361
NM_001289122.1
NP_001276051.1
20, 21
2651
NM_001289124.1
NP_001276053.1
20, 21
2696
NM_014988.3
NP_055803.2
20, 21
2615
XM_005248057.1
XP_005248114.1
27, 28
4247
XM_005248058.1
XP_005248115.1
27, 28
4244
XM_005248060.1
XP_005248117.1
27, 28
4244
XM_005248061.1
XP_005248118.1
26, 27
4163
XM_005248067.2
XP_005248124.1
25, 26
5118
XM_005248072.1
XP_005248129.1
20, 21
2612
XM_005248074.1
XP_005248131.1
19, 20
2576
XM_005248075.1
XP_005248132.1
19, 20
2576
XM_006713996.1
XP_006714059.1
27, 28
4244
XM_011513642.1
XP_011511944.1
28, 29
4322
XM_011513643.1
XP_011511945.1
28, 29
4319
XM_011513644.1
XP_011511946.1
28, 29
4322
XM_011513645.1
XP_011511947.1
27, 28
4238
XM_011513646.1
XP_011511948.1
28, 29
4322
XM_011513648.1
XP_011511950.1
27, 28
4079
XM_011513649.1
XP_011511951.1
23, 24
3981
XM_011513651.2
XP_011511953.1
26, 27
5201
XM_011513653.1
XP_011511955.1
21, 22
2690
XM_011513654.1
XP_011511956.1
20, 21
2654
XM_011513655.1
XP_011511957.1
15, 16
2313
XM_011513656.1
XP_011511958.1
15, 16
2271
XM_011513657.1
XP_011511959.1
14, 15
2235
XM_011513658.2
XP_011511960.1
19, 20
2837
XM_017007894.1
XP_016863383.1
28, 29
4319
XM_017007895.1
XP_016863384.1
27, 28
4247
XM_017007896.1
XP_016863385.1
27, 28
4238
XM_017007897.1
XP_016863386.1
26, 27
4163
XM_017007898.1
XP_016863387.1
20, 21
2615
XM_017007899.1
XP_016863388.1
20, 21
2612
XM_017007900.1
XP_016863389.1
19, 20
2531
XM_017007901.1
XP_016863390.1
19, 20
2531
XM_017007902.1
XP_016863391.1
19, 20
2531
XM_017007903.1
XP_016863392.1
16, 17
2349
XM_017007904.1
XP_016863393.1
15, 16
2271
XM_017007905.1
XP_016863394.1
15, 16
2274
GenBank mRNA
AB029025.2
-
20, 21
2613
AK294774.1
-
14, 15
2424
AK295784.1
-
13, 14
1950
AK295836.1
-
14, 15
2060
AK298915.1
-
15, 16
2267
AK299297.1
-
14, 15
2231
AK299835.1
-
14, 15
2095
AK302674.1
-
7, 8
863
AK309846.1
-
14, 15
2303
AL831962.2
-
25, 26
3999
BC023546.2
-
17, 18
2465
4110496
BC029735.1
-
5, 6
367
BC095394.1
-
10, 11
912
BX537916.1
-
14, 15
2231
BX640692.1
-
13, 14
1989
CR749205.1
-
15, 16
2267
CR933645.1
-
20, 21
2616
CR936601.1
-
20, 21
2622
CR936610.1
-
20, 21
2697
CR936658.1
-
17, 18
2363
CR936661.1
-
19, 20
2516
CR936664.1
-
20, 21
2613
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-nkklt:80/100.66.76.22:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline