Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 3 | 478 | ||
NM_001265611.1 | NP_001252540.1 | 3 | 478 | |||
NM_014994.2 | NP_055809.2 | 3 | 478 | |||
NR_049761.1 | 3 | 478 | ||||
XM_006720438.1 | XP_006720501.1 | 3 | 442 | |||
XM_011521383.1 | XP_011519685.1 | 3 | 442 | |||
XM_011521384.2 | XP_011519686.1 | 3 | 442 | |||
XM_017022017.1 | XP_016877506.1 | 2 | 718 | |||
GenBank mRNA | AB011168.2 | - | 3 | 388 | ||
AK091616.1 | - | 3 | 364 | |||
AK292414.1 | - | 3 | 349 | |||
AL833267.1 | - | 3 | 398 | |||
BC036660.1 | - | 3 | 483 | 5266613 | ||
BC113983.1 | - | 3 | 370 | 40083186 | ||
BC114493.1 | - | 3 | 370 | 40083182 | ||
BX640787.1 | - | 2 | 160 | |||
BX648786.1 | - | 3 | 429 |