Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289003.1 | NP_001275932.1 | 18 | 5612 | ||
NM_015020.3 | NP_055835.2 | 19 | 5466 | |||
GenBank mRNA | AB023148.2 | - | 18 | 5611 | ||
AK001661.1 | - | 1 | 1994 | |||
BX647823.1 | - | 19 | 5466 |