Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308147.1 | NP_001295076.1 | 17 | 4395 | ||
XM_011536624.2 | XP_011534926.1 | 19 | 4676 | |||
XM_011536625.2 | XP_011534927.1 | 19 | 4549 | |||
XM_011536627.2 | XP_011534929.1 | 19 | 4941 | |||
XM_011536628.2 | XP_011534930.1 | 19 | 5127 | |||
XM_011536629.2 | XP_011534931.1 | 18 | 4265 | |||
XM_017021154.1 | XP_016876643.1 | 18 | 4542 | |||
XM_017021155.1 | XP_016876644.1 | 19 | 4631 | |||
XM_017021156.1 | XP_016876645.1 | 19 | 4699 | |||
XM_017021157.1 | XP_016876646.1 | 17 | 4495 | |||
XM_017021158.1 | XP_016876647.1 | 17 | 4134 | |||
GenBank mRNA | AB011171.1 | - | 2 | 2493 | ||
AK074169.1 | - | 11 | 4166 | |||
BC004298.1 | - | 5 | 2205 | |||
BC011891.2 | - | 2 | 2485 | |||
BC063554.1 | - | 3 | 2576 | |||
BC073907.1 | - | 4 | 2683 | |||
BC129952.1 | - | 11 | 3887 | 40126555 | ||
BC129953.1 | - | 17 | 4206 | 40126558 |