Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303614.1 | NP_001290543.1 | 12 | 2326 | ||
NM_018028.3 | NP_060498.2 | 15 | 3047 | |||
XM_011527057.2 | XP_011525359.1 | 13 | 2337 | |||
XM_011527060.2 | XP_011525362.1 | 13 | 3439 | |||
XM_011527061.2 | XP_011525363.1 | 12 | 2251 | |||
XM_011527062.2 | XP_011525364.1 | 12 | 2312 | |||
XM_011527063.2 | XP_011525365.1 | 13 | 2336 | |||
XM_011527064.2 | XP_011525366.1 | 13 | 2337 | |||
XM_017026918.1 | XP_016882407.1 | 12 | 2271 | |||
XM_017026919.1 | XP_016882408.1 | 14 | 2427 | |||
XM_017026920.1 | XP_016882409.1 | 14 | 2449 | |||
XM_017026921.1 | XP_016882410.1 | 12 | 2312 | |||
XM_017026922.1 | XP_016882411.1 | 14 | 2429 | |||
XM_017026923.1 | XP_016882412.1 | 14 | 7967 | |||
GenBank mRNA | AK091041.1 | - | 2 | 124 | ||
BC039652.1 | - | 8 | 1024 | |||
BC054518.1 | - | 15 | 3038 | 6501489 | ||
BC065211.1 | - | 9 | 1334 | |||
BC080641.1 | - | 12 | 2287 | 6645022 | ||
EF601121.1 | - | 15 | 3034 |