Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 4, 5 | 1218 | ||
XM_005263073.3 | XP_005263130.1 | 3, 4 | 1092 | |||
XM_005263075.2 | XP_005263132.1 | 4, 5 | 1257 | |||
XM_005263076.2 | XP_005263133.1 | 4, 5 | 1257 | |||
XM_005263077.3 | XP_005263134.1 | 4, 5 | 1257 | |||
XM_011532037.2 | XP_011530339.1 | 4, 5 | 1270 | |||
XM_017008312.1 | XP_016863801.1 | 5, 6 | 1384 | |||
XM_017008313.1 | XP_016863802.1 | 4, 5 | 1069 | |||
XM_017008314.1 | XP_016863803.1 | 4, 5 | 1068 | |||
XM_017008315.1 | XP_016863804.1 | 4, 5 | 1138 | |||
XM_017008316.1 | XP_016863805.1 | 3, 4 | 1109 | |||
XM_017008317.1 | XP_016863806.1 | 3, 4 | 1081 | |||
XM_017008318.1 | XP_016863807.1 | 4, 5 | 1203 | |||
GenBank mRNA | AJ583824.1 | - | 1, 2 | 152 | ||
BX648339.1 | - | 1, 2 | 272 | |||
CR749490.1 | - | 5, 6 | 1332 |