Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127257.1 | NP_001120729.1 | 10 | 3860 | ||
NM_020342.2 | NP_065075.1 | 10 | 3811 | |||
XM_005246689.4 | XP_005246746.2 | 10 | 3818 | |||
XM_011511504.2 | XP_011509806.1 | 10 | 3725 | |||
XM_011511505.2 | XP_011509807.1 | 11 | 3889 | |||
XM_011511506.2 | XP_011509808.1 | 10 | 6548 | |||
GenBank mRNA | AB033091.1 | - | 10 | 3725 | ||
AK074996.1 | - | 4 | 1671 | |||
BC047493.1 | - | 1 | 151 | |||
BC073909.1 | - | 6 | 2233 | |||
CR749813.1 | - | 10 | 3710 |