Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164313.1 | NP_001157785.1 | 2 | 268 | ||
NM_014797.2 | NP_055612.2 | 2 | 268 | |||
GenBank mRNA | AB007901.1 | - | 2 | 268 | ||
AK309697.1 | - | 2 | 222 | |||
BC036731.1 | - | 2 | 247 | |||
BC117374.1 | - | 1 | 136 | 40125925 | ||
BC117376.1 | - | 1 | 136 | 40125927 |