Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIDINS220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020738.2 | NP_065789.1 | 9 | 1025 | ||
GenBank mRNA | AB033076.2 | - | 9 | 1000 | ||
AK057125.1 | - | 9 | 1013 | |||
AK294832.1 | - | 5 | 329 | |||
AK296459.1 | - | 9 | 1015 | |||
AL133620.1 | - | 9 | 1003 | |||
BC130610.1 | - | 10 | 1120 | 40146641 |