Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304840.1 | NP_001291769.1 | 27 | 4901 | ||
NM_020760.2 | NP_065811.1 | 29 | 5235 | |||
XM_006712646.3 | XP_006712709.1 | 29 | 5293 | |||
XM_006712648.3 | XP_006712711.1 | 28 | 5226 | |||
XM_017004563.1 | XP_016860052.1 | 29 | 5272 | |||
GenBank mRNA | AB037722.1 | - | 29 | 5235 | ||
AL390186.1 | - | 6 | 1000 | |||
CR749424.1 | - | 27 | 4999 |