Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304840.1 | NP_001291769.1 | 16 | 3249 | ||
NM_020760.2 | NP_065811.1 | 18 | 3583 | |||
XM_006712646.3 | XP_006712709.1 | 18 | 3641 | |||
XM_006712648.3 | XP_006712711.1 | 17 | 3574 | |||
XM_017004563.1 | XP_016860052.1 | 18 | 3620 | |||
GenBank mRNA | AB037722.1 | - | 18 | 3583 | ||
BC117194.1 | - | 18 | 3499 | 40125745 | ||
BC117198.1 | - | 18 | 3499 | 40125749 | ||
CR749424.1 | - | 16 | 3347 |